Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129679.2 | NP_181647.1 | 9 - 10 | 1819 | 114 | |
GenBank mRNA | AF153351.1 | - | 9 - 10 | 1527 | 114 | |
AF435448.1 | - | 9 - 10 | 1527 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |