Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIP5K5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129686.2 | NP_181654.1 | 1 - 2 | 1435 | 74 | |
GenBank mRNA | AF260903.1 | - | 1 - 2 | 1396 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |