Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPK20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336999.1 | NP_001324574.1 | 7 - 8 | 1574 | 111 | |
NM_129849.5 | NP_565989.1 | 7 - 8 | 1574 | 111 | ||
GenBank mRNA | AF412082.1 | - | 7 - 8 | 1437 | 111 | |
BT001021.1 | - | 7 - 8 | 1141 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |