Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ML2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084574.1 | NP_001078043.1 | 5 - 6 | 940 | 145 | |
NM_129850.4 | NP_565990.1 | 5 - 6 | 1071 | 145 | ||
NM_201945.3 | NP_973674.1 | 4 - 5 | 1114 | 145 | ||
GenBank mRNA | AF367316.1 | - | 5 - 6 | 897 | 145 | |
AY143940.1 | - | 5 - 6 | 813 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |