Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129863.3 | NP_181830.1 | 5 - 6 | 885 | 67 | |
GenBank mRNA | AF364952.1 | - | 5 - 6 | 803 | 67 | |
BT029025.1 | - | 5 - 6 | 443 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |