Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLO15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129974.1 | NP_181939.1 | 7 - 8 | 738 | 124 | |
NM_201957.2 | NP_973686.1 | 7 - 8 | 884 | 124 | ||
GenBank mRNA | AF369576.1 | - | 7 - 8 | 738 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |