Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEE12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_147236.3 | NP_671769.1 | 2 - 3 | 1603 | 126 | |
GenBank mRNA | AY735572.1 | - | 2 - 3 | 1591 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |