Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAG20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_202550.2 | NP_974279.1 | 1 | 1036 | 89 | |
GenBank mRNA | AY045972.1 | - | 1 | 743 | 90 | |
AY088170.1 | - | 1 | 744 | 90 | ||
BT000768.1 | - | 1 | 745 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |