Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336203.1 | NP_001324547.1 | 8 - 9 | 2763 | 176 | |
NM_128476.3 | NP_180483.1 | 8 - 9 | 3035 | 176 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |