Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP71B29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101180.2 | NP_172769.1 | 1 - 2 | 1006 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |