Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_104651.5 | NP_564746.1 | 2 - 3 | 625 | 100 | |
NM_202320.1 | NP_974049.1 | 2 - 3 | 537 | 100 | ||
GenBank mRNA | AY035134.1 | - | 2 - 3 | 600 | 100 | |
AY113911.1 | - | 1 - 2 | 424 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |