Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGLU22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_105299.3 | NP_176802.1 | 12 - 13 | 1624 | 130 | |
GenBank mRNA | AY074378.1 | - | 12 - 13 | 1415 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |