Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111082.4 | NP_566164.1 | 2 - 3 | 1381 | 115 | |
GenBank mRNA | AY085519.1 | - | 2 - 3 | 926 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |