Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103250.1 | NP_174786.1 | 13 - 14 | 1729 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |