Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AOX1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113134.2 | NP_188875.1 | 3 - 4 | 976 | 146 | |
GenBank mRNA | DQ056603.1 | - | 3 - 4 | 917 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |