Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113259.4 | NP_566730.1 | 5 - 6 | 1330 | 151 | |
GenBank mRNA | AF337954.1 | - | 5 - 6 | 1303 | 151 | |
AY062511.1 | - | 5 - 6 | 1292 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |