Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203796.1 | NP_001190725.1 | 5 - 6 | 3627 | 84 | |
NM_001340902.1 | NP_001319931.1 | 7 - 8 | 3683 | 84 | ||
NM_117491.1 | NP_193150.1 | 6 - 7 | 3549 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |