Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AVA-P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342299.1 | NP_001329512.1 | 2 - 3 | 798 | 140 | |
NM_119638.3 | NP_195198.1 | 2 - 3 | 533 | 140 |
Gene Symbol: AT2G16510 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF372872.1 | - | 2 - 3 | 427 | 140 | |
AY086410.1 | - | 2 - 3 | 495 | 140 | ||
AY127950.1 | - | 2 - 3 | 364 | 140 | ||
L44581.1 | - | 2 - 3 | 411 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |