Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203444.1 | NP_001190373.1 | 1 - 2 | 475 | 173 | |
NM_122291.4 | NP_568437.1 | 1 - 2 | 498 | 173 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |