Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_122737.4 | NP_198206.1 | 2 - 3 | 608 | 103 | |
GenBank mRNA | BT000453.1 | - | 2 - 3 | 391 | 103 | |
BT001212.1 | - | 2 - 3 | 324 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |