Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001344393.1 | NP_001330832.1 | 2 - 3 | 513 | 115 | |
NM_123487.4 | NP_198938.1 | 3 - 4 | 398 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |