Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335093.1 | NP_001325406.1 | 1 - 2 | 727 | 161 | |
NM_001335094.1 | NP_001325405.1 | 2 - 3 | 498 | 161 | ||
NM_001335095.1 | NP_001325407.1 | 2 - 3 | 568 | 161 | ||
NM_126269.3 | NP_178317.2 | 2 - 3 | 342 | 161 | ||
GenBank mRNA | AK117733.1 | - | 2 - 3 | 252 | 161 | |
BT005897.1 | - | 1 - 2 | 188 | 161 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |