Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ProT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336602.1 | NP_001324136.1 | 2 - 3 | 807 | 148 | |
NM_129215.4 | NP_181198.1 | 2 - 3 | 807 | 148 | ||
GenBank mRNA | BT004018.1 | - | 2 - 3 | 396 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |