Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VGD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130272.4 | NP_182227.1 | 1 - 2 | 1236 | 104 | |
GenBank mRNA | AY091768.1 | - | 1 - 2 | 1236 | 104 | |
BT001120.1 | - | 1 - 2 | 1099 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |