Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS28-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340525.1 | NP_001328317.1 | 2 | 1218 | 139 | |
NM_116739.4 | NP_567281.2 | 2 | 967 | 139 | ||
NM_202784.3 | NP_974513.1 | 2 | 732 | 139 | ||
GenBank mRNA | AK229295.1 | - | 2 | 704 | 139 | |
BT015071.1 | - | 2 | 722 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |