Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IPK2BETA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345497.1 | NP_001331991.1 | 2 | 1329 | 81 | |
NM_125570.4 | NP_200984.1 | 2 | 1054 | 81 | ||
GenBank mRNA | AF412073.1 | - | 2 | 1032 | 81 | |
AJ245521.1 | - | 2 | 996 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |