Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128311.3 | NP_180320.1 | 2 | 646 | 156 | |
GenBank mRNA | AY086622.1 | - | 2 | 532 | 156 | |
BT004187.1 | - | 2 | 534 | 156 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |