Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT5G59970 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345379.1 | NP_001332089.1 | 1 | 370 | 87 | |
NM_125390.4 | NP_568918.1 | 1 | 370 | 87 |
Gene Symbol: HIS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY087061.1 | - | 1 | 367 | 87 | |
BT003931.1 | - | 1 | 307 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Both primers and probe map within a single exon |