Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125325.3 | NP_200742.2 | 1 - 2 | 725 | 98 | |
GenBank mRNA | AB493799.1 | - | 1 - 2 | 302 | 98 | |
AY251393.1 | - | 1 - 2 | 365 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |