Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340534.1 | NP_001329797.1 | 6 - 7 | 819 | 159 | |
NM_116751.4 | NP_192421.1 | 7 - 8 | 950 | 159 | ||
GenBank mRNA | AF426400.1 | - | 6 - 7 | 726 | 159 | |
AK175753.1 | - | 7 - 8 | 807 | 159 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |