Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036425.2 | NP_001031502.1 | 2 - 3 | 542 | 97 | |
NM_129283.4 | NP_181264.1 | 3 - 4 | 716 | 97 | ||
GenBank mRNA | AY059849.1 | - | 3 - 4 | 463 | 97 | |
AY081669.1 | - | 2 - 3 | 366 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |