Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336202.1 | NP_001324126.1 | 1 - 2 | 1145 | 139 | |
NM_128471.4 | NP_180478.1 | 2 - 3 | 681 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |