Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035879.1 | NP_001030956.1 | 4 | 440 | 83 | |
NM_100308.4 | NP_171923.1 | 4 | 522 | 83 | ||
GenBank mRNA | AY048221.1 | - | 4 | 443 | 83 | |
AY091703.1 | - | 3 | 352 | 83 | ||
Z23161.1 | - | 381 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |