Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATCSLC08 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335941.1 | NP_001318283.1 | 5 | 2569 | 100 | |
NM_128024.3 | NP_180039.1 | 6 | 2436 | 100 | ||
GenBank mRNA | KJ138900.1 | - | 5 | 1943 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |