Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101170.2 | NP_001094640.2 | 4 - 5 | 572 | 96 | |
GenBank mRNA | BC126593.1 | - | 4 - 5 | 534 | 96 | |
DQ503408.1 | - | 4 - 5 | 572 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |