Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NGB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001853.1 | NP_001001853.1 | 1 - 2 | 90 | 60 | |
GenBank mRNA | AJ635234.2 | - | 1 - 2 | 90 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |