Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZIM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002888.1 | NP_001002888.1 | 3 - 4 | 611 | 71 | |
GenBank mRNA | AY427789.1 | - | 3 - 4 | 611 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Bos taurus Probe spans exons |