Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003905.1 | NP_001003905.1 | 3 - 4 | 515 | 108 | |
GenBank mRNA | AY675080.1 | - | 3 - 4 | 515 | 108 | |
BC112831.1 | - | 3 - 4 | 503 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |