Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007811.2 | NP_001007812.2 | 15 - 16 | 1741 | 65 | |
GenBank mRNA | AY788896.1 | - | 6 - 7 | 661 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Bos taurus Probe spans exons |