Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRABP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008670.1 | NP_001008670.1 | 1 - 2 | 76 | 61 | |
GenBank mRNA | AY821680.1 | - | 1 - 2 | 76 | 61 | |
BC142037.1 | - | 1 - 2 | 203 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |