Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBLN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014946.1 | NP_001014946.1 | 8 - 9 | 1236 | 81 | |
GenBank mRNA | BC105467.1 | - | 8 - 9 | 1225 | 81 | |
BT020707.1 | - | 8 - 9 | 1236 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |