Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034438.1 | NP_001029610.1 | 1 | 185 | 59 | |
GenBank mRNA | BC103289.1 | - | 1 | 185 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon |