Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELAVL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076454.2 | NP_001069922.1 | 4 - 5 | 603 | 81 | |
GenBank mRNA | BC102495.1 | - | 4 - 5 | 604 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |