Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173955.1 | NP_776380.1 | 2 - 3 | 290 | 68 | |
GenBank mRNA | BC102712.1 | - | 2 - 3 | 301 | 68 | |
X52945.1 | - | 290 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |