Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173955.1 | NP_776380.1 | 4 - 5 | 629 | 70 | |
GenBank mRNA | X52945.1 | - | 629 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Bos taurus Probe spans exons |