Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FECH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174054.2 | NP_776479.1 | 3 - 4 | 342 | 66 | |
GenBank mRNA | L34173.1 | - | 3 - 4 | 342 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Bos taurus Probe spans exons |