Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPINF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174140.3 | NP_776565.1 | 2 - 3 | 294 | 55 | |
GenBank mRNA | AF017058.1 | - | 2 - 3 | 296 | 55 | |
BC112656.1 | - | 3 - 4 | 327 | 55 | ||
U48229.1 | - | 2 - 3 | 284 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |