Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174189.3 | NP_776614.2 | 6 - 7 | 881 | 120 | |
GenBank mRNA | BC110213.1 | - | 6 - 7 | 882 | 120 | |
Y17259.1 | - | 881 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |