Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UPK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174214.2 | NP_776639.1 | 2 - 3 | 188 | 85 | |
GenBank mRNA | L20633.1 | - | 2 - 3 | 188 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |