Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174237.2 | NP_776662.1 | 2 - 3 | 408 | 84 | |
GenBank mRNA | BC114157.1 | - | 2 - 3 | 375 | 84 | |
M25757.1 | - | 2 - 3 | 408 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |